Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Acetic acid, Puriss. p.a., ACS Reagent, Reag. ISO, Reag. Ph. Eur., ≥99.8%, Honeywell Fluka™
Shop All Honeywell Research Chemicals Products
Click to view available options
Quantity:
1 L
2.5 L
5 L
30 kg
4 x 2.5 L
60 kg
6 x 1 L
Packaging:
Glass Bottle
Plastic Bottle
Plastic Pail
Description
- Leading the Industry For Over 65 Years
- Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
Specifications
Specifications
| Linear Formula | CH3CO2H |
| UN Number | UN2789 |
| Beilstein | 506007 |
| Formula Weight | 60.05g/mol |
| Percent Purity | ≥99.8% |
| Grade | ACS/puriss. p.a. |
| Density | 1.049g/mL at 25°C |
| Quantity | 60 kg |
| Explosion Limit | 16% at 92°F, 4% at 59°F |
| Flash Point | 40°C (104°F) |
| Show More |
Safety and Handling
| Hazard Statement | P210-P260-P280-P303 + P361 + P353-P305 + P351 + P338-P370 + P378 |
| RTECSNumber | AF1225000 |
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction