Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Acetic Anhydride (Certified ACS), Fisher Chemical™

Catalog No. A10100 Shop All Fisherbrand Products
Click to view available options
Quantity:
100 mL
500 mL
1 L
4 L
Packaging:
Glass Bottle
This item is not returnable. View return policy
This item is not returnable. View return policy

Chemical Identifiers

CAS 108-24-7
Molecular Formula C4H6O3
Molecular Weight (g/mol) 102.089
MDL Number MFCD00008705
InChI Key WFDIJRYMOXRFFG-UHFFFAOYSA-N
Synonym acetic anhydride, acetanhydride, acetic acid, anhydride, acetic oxide, acetyl oxide, ethanoic anhydride, acetyl ether, acetyl anhydride, acetic acid anhydride, anhydride acetique
PubChem CID 7918
ChEBI CHEBI:36610
IUPAC Name acetyl acetate
SMILES CC(=O)OC(=O)C

Specifications

Melting Point -73.1°C
Color Colorless
Density 1.087g/cm³
pH 3
Boiling Point 140°C
Flash Point 53°C
Assay Percent Range ≥97.0 %
Packaging Glass Bottle
Linear Formula (CH3CO)2O
Quantity 100 mL
UN Number 1715
Residue after Evaporation 0.002% max.
Formula Weight 102.09
Percent Purity ≥97.0%
Grade Certified ACS
Physical Form Liquid
Chemical Name or Material Acetic Anhydride
Show More Show Less
DOTInformation DOT Class 8, : Corrosive
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.