Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Acetic Anhydride, Reagent, ACS, 97%, Spectrum™ Chemical

Click to view available options
Quantity:
125 mL
500 mL
2.5 L
4 L
6 x 500 mL
20 L
4 x 2.5 L
4 x 4 L
200 L
Packaging:
Amber Glass Bottle
Drum
Glass Bottle
Poly Drum
Description
Spectrum™ Chemical Acetic Anhydride, Reagent, ACS , or ethanoic anhydride as it is otherwise known is a colorless, strong smelling liquid chemical compound that is the simplest isolatable acid anhydride and commonly used as a reagent for organic synthesis. Mainly used for acetylations, its main use is for the conversion of cellulose to cellulose acetate and in the synthesis of aspirin. As an ACS grade Reagent, Spectrum Chemical manufactured acetic anhydride is used as the quality standard against which other substances are graded and has met the toughest regulatory standards for quality and pureness.
Specifications
Specifications
CAS Max % | 100% |
Packaging | Glass Bottle |
Quantity | 2.5 L |
UN Number | UN1715 |
Residue after Evaporation | 0.003% |
Percent Purity | 97% |
Grade | ACS |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction