Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Acetoacetyl CoA synthetase Antibody, DyLight 405, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP297174V
Description
Acetoacetyl CoA synthetase Polyclonal specifically detects Acetoacetyl CoA synthetase in Human samples. It is validated for Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.Specifications
Acetoacetyl CoA synthetase | |
Polyclonal | |
50 mM Sodium Borate | |
Rabbit | |
Antigen Affinity-purified | |
RUO | |
65985 | |
Store at 4°C in the dark. | |
IgG |
Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
DyLight 405 | |
acetoacetate-CoA ligase, acetoacetyl-CoA synthetase, ACSF1FLJ41251, Acyl-CoA synthetase family member 1, EC 6.2.1, EC 6.2.1.16, FLJ12389, homolog of C. elegans supressor of ras 5 (sur-5), Protein sur-5 homolog, SUR-5 | |
Produced in rabbits immunized with E. coli-derived Human Acetoacetyl CoA synthetase fragment. | |
0.1 mL | |
Primary | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction