Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Acetoacetyl coenzyme A sodium salt, >90%, MP Biomedicals™

Catalog No. ICN15022410
Click to view available options
Quantity:
1 mg
10 mg
25 mg
50 mg

Chemical Identifiers

CAS 102029-52-7
Molecular Formula C25H40N7NaO18P3S
Molecular Weight (g/mol) 874.597
InChI Key RSOOKMDOLPTBRH-UHFFFAOYSA-N
PubChem CID 133112705
IUPAC Name S-[2-[3-[[4-[[[5-(6-aminopurin-9-yl)-4-hydroxy-3-phosphonooxyoxolan-2-yl]methoxy-hydroxyphosphoryl]oxy-hydroxyphosphoryl]oxy-2-hydroxy-3,3-dimethylbutanoyl]amino]propanoylamino]ethyl] 3-oxobutanethioate;sodium
SMILES CC(=O)CC(=O)SCCNC(=O)CCNC(=O)C(C(C)(C)COP(=O)(O)OP(=O)(O)OCC1C(C(C(O1)N2C=NC3=C2N=CN=C3N)O)OP(=O)(O)O)O.[Na]

Specifications

Color Yellow
Assay Percent Range >90%
Quantity 10 mg
Formula Weight 901.6 (free acid)
Physical Form Solid
Chemical Name or Material Acetoacetyl coenzyme A
Recommended Storage Store at -20°C.
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.