Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Acetoin, 96%, dimer

Catalog No. ac411951000
Click to view available options
Quantity:
100 g
500 g
Packaging:
Glass bottle
This item is not returnable. View return policy
This item is not returnable. View return policy

CAS: 23147-57-1 | C8H16O4 | 176.21 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 23147-57-1
Molecular Formula C8H16O4
Molecular Weight (g/mol) 176.21
MDL Number MFCD00038696
InChI Key DFMGATPNJMFDCR-UHFFFAOYNA-N
Synonym acetoin, 3-hydroxy-2-butanone, acetyl methyl carbinol, dimethylketol, 2-butanone, 3-hydroxy, acetylmethylcarbinol, 2,3-butanolone, 2-hydroxy-3-butanone, 1-hydroxyethyl methyl ketone, methanol, acetylmethyl
PubChem CID 179
ChEBI CHEBI:15688
IUPAC Name 3-hydroxybutan-2-one
SMILES CC1OC(C)(O)C(C)OC1(C)O

Specifications

CAS Min % 95.0
CAS Max % 100.0
Melting Point 15.0°C
Color Green-Yellow to Yellow or White to Yellow
Density 1.0000g/mL
Boiling Point 148.0°C
Flash Point 50°C
Infrared Spectrum Authentic
Assay Percent Range 95% min. (GC)
Packaging Glass bottle
Refractive Index 1.4171
Linear Formula CH3COCH(OH)CH3
Quantity 100 g
Beilstein 01, 827
Specific Gravity 1
Merck Index 15, 63
Solubility Information Solubility in water: soluble. Other solubilities: soluble in alcohol,propylene glycol,insoluble in vegetable oil
Formula Weight 88.11
Percent Purity 96%
Physical Form Liquid (Monomer) or Powder or Crystals (Dimer)
Chemical Name or Material Acetoin, pract., 93%
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement
Flammable liquid and vapor.
Causes skin irritation.
Causes serious eye irritation.
Precautionary Statement GHS P Statement
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
IF ON SKIN: Wash with plenty of soap and water.
Take off contaminated clothing and wash before reuse.
IF IN EYES: Rinse cautiously with water for
EINECSNumber 208-174-1
RTECSNumber EL8790000
TSCA TSCA
Recommended Storage may polymerize during storage

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.