Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Acetone, 99+%, extra pure

Catalog No. AC177170050
Click to view available options
Quantity:
1 L
2.5 L
5 L
10 L
25 L
Packaging:
Glass Bottle
Plastic Drum

CAS: 67-64-1 | C3H6O | 58.08 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 67-64-1
Molecular Formula C3H6O
Molecular Weight (g/mol) 58.08
MDL Number MFCD00008765
InChI Key CSCPPACGZOOCGX-UHFFFAOYSA-N
Synonym acetone, 2-propanone, propanone, dimethyl ketone, methyl ketone, dimethylformaldehyde, pyroacetic ether, beta-ketopropane, dimethylketal, chevron acetone
PubChem CID 180
ChEBI CHEBI:15347
IUPAC Name propan-2-one
SMILES CC(C)=O

Specifications

Melting Point -95.0°C
CAS Min % 99.0
CAS Max % 100.0
Decomposition Information >4°C
Density 0.7900g/mL
Boiling Point 56.0°C
Quantity 5 L
Flash Point −20°C
Packaging Plastic Drum
Physical Form Liquid
Refractive Index 1.3585
Residue after Evaporation 0.005% max.
Assay Percent Range 99+%
Specific Gravity 0.79
Linear Formula CH3COCH3
Vapor Pressure Vapor Pressure: 247mbar at 20°C
Viscosity 0.32 mPa.s (20°C)
UN Number 1090; Colorless
Beilstein 01, 635
Water 0.5% max. (K.F.)
Fieser 02,13; 03,4; 06,9
Merck Index 15, 65
Solubility Information Solubility in water: soluble. Other solubilities: miscible with alcohol, dimethylformamide,, chloroform, ether and most oils
Formula Weight 58.08
Percent Purity ≥99%
Grade Extra Pure
Chemical Name or Material Acetone
Show More Show Less
Hazard Category Danger
Hazard Statement GHS H Statement:
May cause drowsiness or dizziness.
Causes serious eye irritation.
Highly flammable liquid and vapor.
Repeated exposure may cause skin dryness or cracking.
Precautionary Statement GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Ground/Bond container and receiving equipment.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.