Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Lead(IV) acetate, 95%, stabilized
Click to view available options
Quantity:
100 g
500 g
2.5 kg
Packaging:
Glass bottle
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Oxidation reagent

Specifications
Specifications
Melting Point | 175.0°C to 180.0°C |
Color | Orange-Pink to White |
Physical Form | Crystalline Powder |
Quantity | 500 g |
Assay Percent Range | 94% min. (Iodometry) |
Linear Formula | (CH3CO2)4Pb |
Fieser | 01,537; 02,234; 03,168; 04,278; 05,365; 06,313; 08,269; 09,265; 10,228; 12,270; 13,62; 14,188; 16,193 |
Merck Index | 15,5478 |
Solubility Information | Solubility in water: decomposes in water forming pb02 and acetic acid. Other solubilities: soluble in hot glacial acetic acid, soluble in benzene, chloroform, tetrachloroethane,, toluene, acetonitrile, nitrobenzene and, dichloromethane |
Formula Weight | 443.36 |
Show More |
Safety and Handling
Hazard Category | GHS Signal Word: Danger |
Hazard Statement | GHS H Statement May damage the unborn child. Suspected of damaging fertility. Harmful if inhaled. Harmful if swallowed. May cause damage to organs through prolonged or repeated exposure. Very toxic to aquatic life with |
Precautionary Statement | GHS P Statement Use personal protective equipment as required. Do not breathe dust/fume/gas/mist/vapors/spray. Avoid release to the environment. IF exposed or concerned: Get medical advice/attention. |
EINECSNumber | 208-908- |
RUO – Research Use Only
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction