Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Acetone/Acetonitrile, 80/20, HPLC Grade, Ricca Chemical
SDP

Catalog No. R019500020E Shop All Ricca Chemical Company Products
Change view
Click to view available options
Quantity:
4 L
20 L
Packaging:
Amber Glass Bottle
Metal Can
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
R019500020E 20 L Metal Can
R01950004C 4 L Amber Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. R019500020E Supplier Ricca Chemical Company Supplier No. R019500020E4
Add to Cart
Add to Cart

Chemical Identifiers

CAS 67-64-1 , 75-05-8
Molecular Formula C3H6O
Molecular Weight (g/mol) 58.08
MDL Number MFCD00008765
InChI Key CSCPPACGZOOCGX-UHFFFAOYSA-N
Synonym acetone, 2-propanone, propanone, dimethyl ketone, methyl ketone, dimethylformaldehyde, pyroacetic ether, beta-ketopropane, dimethylketal, chevron acetone
PubChem CID 180
ChEBI CHEBI:15347
IUPAC Name propan-2-one
SMILES CC(C)=O

Specifications

Color Colorless
CAS Min % 78.0, 19.59
CAS Max % 82.0, 20.39
Quantity 20 L
Packaging Metal Can
Physical Form Liquid
Boiling Point 56°C (Acetone), 82°C (Acetonitrile)
Percent Purity 80/20
Grade HPLC
Chemical Name or Material Acetone/Acetonitrile, 80 (Acetone) / 20 (Acetonitrile)
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.