Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Acetone, MP Biomedicals™

Catalog No. p-4405641 Shop All MP Biomedicals, Inc Products
Click to view available options
Quantity:
100 mL
0.5 L
1 L
4 L

Acetone ACS, ≥99.5%, Clear, Colorless Liquid

  • It is a commonly used material in the manufacture of various chemicals like methyl isobutyl ketone, mesityl oxide, diacetone alcohol, acetic acid (via the ketene process), iodoform, and bromoform.
  • Other products where acetone is utilized in manufacture include rayon, photographic films, and paint and varnish removers.
  • In chemical synthesis, acetone has been utilized in the synthesis and isolation of both organic and inorganic compounds and complexes.
  • These include t-butanesulfinamide, terpenes, thiol-stabilized gold colloids, calixphyrin macrocycles, and trispyrazolylborate platinum compounds.
  • Acetone is also used in HPLC analysis of organic and inorganic species, including gold nanoparticle clusters and plant oils. Acetone has been utilized in protein crystallization.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 67-64-1
Molecular Formula C3H6O
Molecular Weight (g/mol) 58.08
MDL Number MFCD00008765
InChI Key CSCPPACGZOOCGX-UHFFFAOYSA-N
PubChem CID 180
ChEBI CHEBI:15347
IUPAC Name propan-2-one
SMILES CC(C)=O

Specifications

Melting Point -95.4°C D396(Literature)
Color ≤10 (APHA)
Density 0.7905 - 0.793g/cm3(Lit.)
Boiling Point 56.2°C (at 1013 hPa) (literature)
Quantity 100 mL
Flash Point -17°C / 1.4°F
Physical Form Liquid
Refractive Index n20/D 1.359(lit.)
Assay Percent Range ≥99.5%
Titratable Base ≤0.0006 meq/g
Vapor Pressure 184 mmHg (20°C)
UN Number UN1090
Water ≤0.5%
Beilstein 635680EC
Synonym acetone, 2-propanone, propanone, dimethyl ketone, methyl ketone, dimethylformaldehyde, pyroacetic ether, beta-ketopropane, dimethylketal, chevron acetone
Solubility Information Soluble in water.
Formula Weight 58.1
Grade ≥99.5%
Chemical Name or Material Acetone, For ACS analysis
Show More Show Less
RTECSNumber AL3150000
storageNote1 Store at Room Temperature(15- 30°C).
WARNING: Cancer and Reproductive Harm - www.P65Warnings.ca.gov
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.