Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Acetone, BAKER ANALYZED™ A.C.S. Reagent, J.T. Baker™

Catalog No. 02002158
Click to view available options
Quantity:
500 mL
1 L
4 L
Packaging:
Aluminum Bottle
HDPE Pressure Bottom Poly Bottle
Narrow Mouth Amber Glass Bottle
Narrow Mouth PVC-coated Amber Glass Bottle
This item is not returnable. View return policy
This item is not returnable. View return policy

High quality chemicals for laboratory and specialized industrial use

    For Laboratory,Research,or Manufacturing Use

Chemical Identifiers

CAS 67-64-1
Molecular Formula C3H6O
Molecular Weight (g/mol) 58.08
MDL Number MFCD00008765
InChI Key CSCPPACGZOOCGX-UHFFFAOYSA-N
Synonym acetone, 2-propanone, propanone, dimethyl ketone, methyl ketone, dimethylformaldehyde, pyroacetic ether, beta-ketopropane, dimethylketal, chevron acetone
PubChem CID 180
ChEBI CHEBI:15347
IUPAC Name propan-2-one
SMILES CC(C)=O

Specifications

Boiling Point ≤56.1°C
Quantity 500 mL
Flash Point -20°C (-4°C)
Packaging HDPE Pressure Bottom Poly Bottle
Specific Gravity 1L = 0.79kg
Formula Weight 58.08
Grade Meets ACS reagent chemical requirements
Chemical Name or Material Acetone
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.