Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Acetone cyanohydrin, 99%, stabilized

Catalog No. ac102380050
Click to view available options
Quantity:
5 g
Packaging:
Glass bottle

CAS: 75-86-5 | C4H7NO | 85.11 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 75-86-5
Molecular Formula C4H7NO
Molecular Weight (g/mol) 85.11
InChI Key MWFMGBPGAXYFAR-UHFFFAOYSA-N
Synonym acetone cyanohydrin, acetone cyanhydrin, 2-methyllactonitrile, 2-hydroxyisobutyronitrile, alpha-hydroxyisobutyronitrile, acetoncyanhydrin, 2-cyano-2-propanol, acetoncianidrina, acetonkyanhydrin, acetoncianhidrinei
PubChem CID 6406
ChEBI CHEBI:15348
IUPAC Name 2-hydroxy-2-methylpropanenitrile
SMILES CC(C)(C#N)O

Specifications

Melting Point -19.0°C
Color Colorless to Yellow
Density 0.9320g/mL
Boiling Point 82.0°C (23.0 mmHg)
Flash Point 73°C
Infrared Spectrum Authentic
Assay Percent Range 98.5% min. (GC)
Packaging Glass bottle
Refractive Index 1.3990 to 1.4010
Linear Formula (CH3)2C(OH)CN
Quantity 5 g
Beilstein 03,316
Fieser 01,5; 08,409; 11,1
Merck Index 15,66
Specific Gravity 0.932
Solubility Information Solubility in water: soluble.
Stabilizer Approx. 0.3% H3PO4
Viscosity 4.8 mPa.s (20°C)
Formula Weight 85.11
Percent Purity 99%
Physical Form Liquid
Chemical Name or Material Acetone cyanohydrin
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement
Fatal if swallowed.
Fatal in contact with skin.
Fatal if inhaled.
Very toxic to aquatic life with long lasting effects.
Precautionary Statement GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
Immediately call a POISON CENTRE or doctor/physician.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if pre
EINECSNumber 200-909-4

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.