Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Acetone, for gas chromatography MS SupraSolv™, MilliporeSigma™

Catalog No. p-7020408
Click to view available options
Quantity:
1 L
2.5 L
Packaging:
Glass Bottle

For gas chromatography MS SupraSolv™

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 67-64-1
Molecular Formula C3H6O
Molecular Weight (g/mol) 58.08
MDL Number MFCD00008765
InChI Key CSCPPACGZOOCGX-UHFFFAOYSA-N
Synonym acetone, 2-propanone, propanone, dimethyl ketone, methyl ketone, dimethylformaldehyde, pyroacetic ether, beta-ketopropane, dimethylketal, chevron acetone
PubChem CID 180
ChEBI CHEBI:15347
IUPAC Name propan-2-one
SMILES CC(C)=O

Specifications

pH 5 to 6 (at 395g/L 20°C)
Melting Point -95.4°C
Color Colorless
Density 0.79g/cm3 at 20°C
Boiling Point 56.2°C
Quantity 1 L
Flash Point <−20°C (<−4°F)
Ignition Point 465°C (869°F)
Odor Fruity
Packaging Glass Bottle
Physical Form Liquid
Refractive Index 1.35
Vapor Pressure 233 hPa (20°C)
UN Number 1090
Solubility Information Soluble in water at 20°C
Formula Weight 58.08
Chemical Name or Material Acetone
Show More Show Less
Hazard Category H225: Highly flammable liquid and vapour.
Hazard Statement H319: Causes serious eye irritation.
Precautionary Statement H336 May cause drowsiness and dizziness.
Recommended Storage 5°C to 30°C (41°F to 86°F); Keep container tightly closed in a dry and well-ventilated place. Keep away from heat and sources of ignition. Keep locked up or in an area accessible only to qualified or authorized persons.
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.