Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Acetone, for Spectroscopy Uvasol™, MilliporeSigma™
Shop All MilliporeSigma Products
Click to view available options
Quantity:
500 mL
Packaging:
Glass Bottle
Specifications
Specifications
pH | 5 to 6 (at 395g/L 20°C) |
Melting Point | -95.4°C |
Color | Colorless |
Density | 0.79g/cm3 at 20°C |
Boiling Point | 56.2°C |
Quantity | 500 mL |
Flash Point | <−20°C (<−4°F) |
Ignition Point | 465°C (869°F) |
Odor | Fruity |
Packaging | Glass Bottle |
Show More |
Safety and Handling
Hazard Category | H225: Highly flammable liquid and vapour. |
Hazard Statement | H319: Causes serious eye irritation. |
Precautionary Statement | H336 May cause drowsiness and dizziness. |
Recommended Storage | 15° to 25°C (59° to 77°F); Keep container tightly closed. Keep container in a cool, well-ventilated place. Keep away from heat and source of ignition. Protected from light. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction