Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Acetone, N.F. - F.C.C., J.T.Baker™

Click to view available options
Quantity:
500mL
4L
Packaging:
Narrow Mouth Amber Glass Bottle
67-64-1
C3H6O
58.08
CSCPPACGZOOCGX-UHFFFAOYSA-N
acetone,2-propanone,propanone,dimethyl ketone,methyl ketone,dimethylformaldehyde,pyroacetic ether,beta-ketopropane,dimethylketal,chevron acetone
180
CHEBI:15347
propan-2-one
CC(=O)C
67-64-1
C3H6O
58.08
CSCPPACGZOOCGX-UHFFFAOYSA-N
acetone,2-propanone,propanone,dimethyl ketone,methyl ketone,dimethylformaldehyde,pyroacetic ether,beta-ketopropane,dimethylketal,chevron acetone
180
CHEBI:15347
propan-2-one
CC(=O)C
Formula Weight 58.08
Grade FCC/NF
Packaging Narrow Mouth Amber Glass Bottle
Specific Gravity 1L = 0.79kg
Flash Point −20.00°C
Quantity 4L
Chemical Name or Material Acetone