Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Acetone oxime, 98%
Click to view available options
Quantity:
50 g
250 g
1000 g
Description
It is an intermediate used in organic synthesis and in agriculture. It is also an important raw material.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
It is an intermediate used in organic synthesis and in agriculture. It is also an important raw material.
Solubility
Soluble in water (330 g/L at 20°C).
Notes
Store away from oxidizing agents. Protect against electrostatic charges.
It is an intermediate used in organic synthesis and in agriculture. It is also an important raw material.
Solubility
Soluble in water (330 g/L at 20°C).
Notes
Store away from oxidizing agents. Protect against electrostatic charges.

Specifications
Specifications
Melting Point | 60°C to 63°C |
Density | 0.901 |
Boiling Point | 133°C to 135°C |
Flash Point | 60°C (140°F) |
Quantity | 1000 g |
UN Number | UN1325 |
Beilstein | 1560146 |
Merck Index | 14,75 |
Solubility Information | Soluble in water (330g/L at 20°C). |
Formula Weight | 73.1 |
Show More |
Safety and Handling
Hazard Category | H228-H318-H373 |
Hazard Statement | GHS H Statement H228 Flammable solid. |
Precautionary Statement | P210-P240-P241-P260-P280-P305+P351+P338-P310-P314-P370+P378q-P501c |
DOTInformation | Hazard Class: 4.1; Packaging Group: II |
EINECSNumber | 204-820-1 |
RTECSNumber | AL6825000 |
TSCA | Yes |
Recommended Storage | Ambient temperatures |
RUO – Research Use Only
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction