Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Acetone oxime, 98%

Catalog No. p-48436
Click to view available options
Quantity:
100 g
500 g
2.5 kg
Packaging:
Glass bottle
Plastic Bottle

CAS: 127-06-0 | C3H7NO | 73.10 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 127-06-0
Molecular Formula C3H7NO
Molecular Weight (g/mol) 73.10
MDL Number MFCD00002118
InChI Key PXAJQJMDEXJWFB-UHFFFAOYSA-N
Synonym acetone oxime, acetoxime, propan-2-one oxime, 2-propanone, oxime, 2-propanone oxime, acetonoxime, acetone, oxime, beta-isonitrosopropane, acetoneoxime, ccris 5
PubChem CID 67180
ChEBI CHEBI:15349
IUPAC Name N-propan-2-ylidenehydroxylamine
SMILES CC(C)=NO

Specifications

CAS Min % 97.5
CAS Max % 100.0
Melting Point 58.0°C to 62.0°C
Color White to Yellow
Boiling Point 135.0°C
Flash Point 60°C
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (GC)
Packaging Glass bottle
Linear Formula (CH3)2C=NOH
Quantity 100 g
Beilstein 01, 649
Fieser 13,22
Merck Index 15, 73
Solubility Information Solubility in water: 330g/L (20°C). Other solubilities: soluble in alcohols and ethers
Formula Weight 73.09
Percent Purity 98%
Physical Form Crystalline Solid or Crystals
Chemical Name or Material Acetone oxime, 98%
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement
Flammable solid.
Causes serious eye damage.
May cause damage to organs through prolonged or repeated exposure.
Precautionary Statement GHS P Statement
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsing.
Immediately call
EINECSNumber 204-820-1
RTECSNumber AL6825000
TSCA TSCA

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.