Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Acetone Pharmaceutical Secondary Standard, Certified Reference Material, MilliporeSigma™ Supelco™

Catalog No. 111019582 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Quantity:
3 x 1.5 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
11-101-9582 3 x 1.5 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 11-101-9582 Supplier MilliporeSigma Supelco Supplier No. PHR12193X1.5ML
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Pharmaceutical secondary standards for application in quality control provide pharma laboratories and manufacturers with a convenient and cost-effective alternative to the preparation of in-house working standards. Acetone is an organic compound and in contrast to petroleum based hexane, it is a biorenewable solvent obtained via fermentation. It is generally used as a solvent for many gums, resins, and in the production of plastics.

This Certified Reference Material (CRM) is produced and certified in accordance with ISO 17034 and ISO/IEC 17025. All information regarding the use of this CRM can be found on the certificate of analysis.
TRUSTED_SUSTAINABILITY

Specifications

CAS 67-64-1
Vapor Pressure 184 mmHg (20°C)
Vapor Density 2 (vs. air)
Density 0.791 g/mL (at 25°C)
Physical Form Neat
Linear Formula CH3COCH3
MDL Number MFCD00008765
Melting Point -94°C
Boiling Point 56°C (760 mmHg)
Quantity 3 x 1.5 mL
Formula Weight 58.08
Grade Certified Reference Material
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.