Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Acetone, Puriss., meets analytical spec. of BP, NF, Ph. Eur., ≥99% (GC), Honeywell Riedel-de Haën™
Click to view available options
Quantity:
1 L
2.5 L
5 L
20 kg
4 x 2.5 L
4 x 5 L
6 x 1 L
155 kg
Packaging:
Glass Bottle
Metal Drum
Poly Bottle
Stainless Steel Can
Description
- Honeywell Riedel-de Haen has over 200 years of history in producing high-purity chemicals
- It is the leading brand offering High-Purity Solvents for dedicated applications.

Specifications
Specifications
Melting Point | -95°C |
Color | Colorless |
Density | 0.791g/mL at 25°C, 0.790 to 0.792g/mL at 20°C |
Boiling Point | 55°C to 57°C |
Quantity | 2.5 L |
Explosion Limit | 0.132 |
Flash Point | −17°C (1.4°F) |
Packaging | Glass Bottle |
Physical Form | Liquid |
Linear Formula | CH3COCH3 |
Show More |
Safety and Handling
Hazard Statement | P210-P305 + P351 + P338-P370 + P378-P403 + P235 |
Precautionary Statement | H225-H319-H336 |
DOTInformation | Class 3 - PG 2 - Acetone |
EINECSNumber | 200-662-2 |
RTECSNumber | AL3150000 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction