Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Acetone, Semiconductor Grade, 99.5%

Catalog No. AA19392K7
Click to view available options
Quantity:
1 L
4 L
4 x 1 L

Maximum level of impurities: Color (APHA) 10, Evaporation residue 5ppm, Acidity 0.3μeq/g, Alkalinity 0.5μeq/g, H2O 0.5%, Solubility in H2O P.T., Cl 0.2ppm, PO4 0.1ppm | CAS: 67-64-1 | C3H6O | 58.08 g/mol

Acetone, semiconductor grade is suitable for use in electronics and circuit board manufacturing. Its luminescence intensity is dependent upon the solution components and the absorption of UV light induces photolysis and production of radials.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 67-64-1
Molecular Formula C3H6O
Molecular Weight (g/mol) 58.08
MDL Number MFCD00008765
InChI Key CSCPPACGZOOCGX-UHFFFAOYSA-N
Synonym acetone, 2-propanone, propanone, dimethyl ketone, methyl ketone, dimethylformaldehyde, pyroacetic ether, beta-ketopropane, dimethylketal, chevron acetone
PubChem CID 180
ChEBI CHEBI:15347
IUPAC Name propan-2-one
SMILES CC(C)=O

Specifications

Melting Point 94°C
Color Colorless
Appearance Colorless liquid
Density 0.791 g/cm3
Boiling Point 56°C
Quantity 4 L
Flash Point -17°C (1°F)
Ignition Point 465°C (869°F)
Odor Characteristic
Physical Form Liquid
Refractive Index 1.359
Assay Percent Range 99.5%
Vapor Pressure 247 hPa (185mm Hg) at 20°C
Beilstein 63580
Solubility Information Miscible with water,benzene,alcohol,dimethylformamide,ether,chloroform and most oils.
Formula Weight 58.08
Grade Semiconductor
Chemical Name or Material Acetone
Show More Show Less
Hazard Category H225-H319-H336-H373
Precautionary Statement P210-P233-P235-P240-P241-P242-P243-P260-P264b-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P312-P314-P370+P378q-P501c
DOTInformation Hazard Class: 3, Packing Group: II
EINECSNumber 200-662-2
RTECSNumber AL3150000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.