Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Acetonitrile, 80%, 20% Water with 0.1% Formic Acid, Optima LC/MS, Fisher Chemical

Catalog No. NC0959509 Shop All Fisherbrand Products
Encompass
Click to view available options
Quantity:
500 mL
  • Reduced concentration of acetonitrile improves chromatographic performance, especially of early eluting, hydrophilic peptides.
  • Pre-mixed for ease-of-use.
  • Low metal ion content to ensure minimal metal contamination.
  • Ultra filtered to ensure low levels of particulate contamination.
  • Recommended solvent for use with Thermo Scientific EASY-nLC 1200 and Ultimate RSLCnano.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 75-05-8, 7732-18-5, 64-18-6
Molecular Formula C2H3N
Molecular Weight (g/mol) 41.053
InChI Key WEVYAHXRMPXWCK-UHFFFAOYSA-N
Synonym methyl cyanide, cyanomethane, ethanenitrile, methanecarbonitrile, ethyl nitrile, methane, cyano, acetonitrile, cyanure de methyl, methylkyanid, mecn
PubChem CID 6342
ChEBI CHEBI:38472
IUPAC Name acetonitrile
SMILES CC#N

Specifications

Quantity 500 mL
Odor Aromatic
Physical Form Liquid
Specific Gravity 0.7810
Chemical Name or Material Acetonitile with formic acid
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.