Learn More
Acetonitrile 90% and Water 10% with 0.1% formic acid, LabReady™ Solvent Blend, for LC-MS, for HPLC, Solstice
Shop All Solstice Research Chemicals ProductsDescription
LabReady™ Solvent Blends
- Using custom blends not only helps you reduce waste through human error, but also cuts disposal costs for hazardous materials used during blend preparation
- Solstice Research Chemicals uses proprietary, closed-loop blending technology, which eliminates variation, ensuring your blends are as precise as possible
- Solstice LabReady Blends undergo full safety analyses, with a safety data sheet provided for every blended product
- Solstice's high-precision blending technology reduces contamination and exposure during the blending process, helping to minimize environmental risk
Specifications
Specifications
| CAS Max % | 87.6, 6.3, <0.20 |
| Density | 0.8036 g/cm3 |
| Quantity | 4 x 2.5 L |
| Flash Point | 8.33°C |
| Packaging | Glass Bottle |
| Physical Form | Liquid |
| Boiling Point | 66.1°C |
| Grade | LC-MS |
Safety and Handling
| Hazard Category | Danger |
| Hazard Statement | H370+H319+H302+H332+H225 |
| Precautionary Statement | P280+P210+P233+P240+P241+P242+P243+P260+P264+P270+P271 |
| Recommended Storage | Room Temperature |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.