Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Acetonitrile, For pesticide residue analysis, Solstice

Catalog No. 6000258 Shop All Honeywell Research Chemicals Products
Change view
Click to view available options
Quantity:
1 L
2.5 L
4 x 2.5 L
4 x 4 L
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
60-002-58 1 L
60-002-59 2.5 L
60-046-343 4 x 2.5 L
60-048-229 4 x 4 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 60-002-58 Supplier Solstice Supplier No. 344811L
Add to Cart
Add to Cart

for pesticide residue analysis

  • For gas chromatography (GC)
  • Developed especially for the application in residue analysis of pesticides and other low-volatile substances using GC
  • Non-volatile matter and water content are specified for each solvent
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 75-05-8
Molecular Formula C2H3N
MDL Number MFCD00001878
InChI Key WEVYAHXRMPXWCK-UHFFFAOYSA-N
PubChem CID 6342
ChEBI CHEBI:38472
IUPAC Name acetonitrile
SMILES CC#N

Specifications

Color Colorless
Density 0.786g/mL at 25°C
Quantity 1 L
Explosion Limit 0.16
Flash Point 2°C (35.6°F)
Ignition Point 973°F
Linear Formula CH3CN
Vapor Pressure 72.8 mmHg (20°C)
UN Number UN1648
Beilstein 741857
Synonym methyl cyanide, cyanomethane, ethanenitrile, methanecarbonitrile, ethyl nitrile, methane, cyano, acetonitrile, cyanure de methyl, methylkyanid, mecn
Molecular Weight (g/mol) 41.053
Formula Weight 41.05g/mol
Percent Purity ≥99.9% (GC)
Grade CHROMASOLV
Chemical Name or Material Acetonitrile
Show More Show Less
Hazard Statement P210-P280-P305 + P351 + P338
RTECSNumber AL7700000
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.