Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Acetonitrile, AMD CHROMASOLV™, ≥99.9%, Solstice

Catalog No. 6000230 Shop All Honeywell Research Chemicals Products
Change view
Click to view available options
Quantity:
1 L
6 x 1 L
Packaging:
Glass Bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
60-002-30 1 L Glass Bottle
60-046-502 6 x 1 L Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 60-002-30 Supplier Solstice Supplier No. 348961L
Add to Cart
Add to Cart

CHROMASOLV™ AMD,≥99.9%

  • Solvents designed for high-performance thin-layer chromatography
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 75-05-8
Molecular Formula C2H3N
Molecular Weight (g/mol) 41.053
MDL Number MFCD00001878
InChI Key WEVYAHXRMPXWCK-UHFFFAOYSA-N
Synonym methyl cyanide, cyanomethane, ethanenitrile, methanecarbonitrile, ethyl nitrile, methane, cyano, acetonitrile, cyanure de methyl, methylkyanid, mecn
PubChem CID 6342
ChEBI CHEBI:38472
IUPAC Name acetonitrile
SMILES CC#N

Specifications

Melting Point -48°C (literature)
CAS Max % 100.0000%
Density 0.786 g/mL at 25 °C(lit.)
Quantity 1 L
Flash Point 6°C
Packaging Glass Bottle
Linear Formula CH3CN
UN Number UN1648
Beilstein 741857
Boiling Point 81.6°C
Formula Weight 41.05
Percent Purity >99.9%
Grade High Purity
Chemical Name or Material Acetonitrile, AMD CHROMASOLV™, ≥99.9%
Show More Show Less
ShelfLife 1260 days from date of manufacture
DOTInformation Transport Hazard Class: 3; Packing Group:3: II; Proper Shipping Name: Acetonitrile
EINECSNumber 200-835-2
Recommended Storage Room Temp
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.