Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Acetonitrile, Anhydrous, For Biotechnology, MilliporeSigma™

Catalog No. MAX01521 Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
4 L
20 L
Packaging:
Plastic Barrel
Plastic Bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
MAX01521 4 L Plastic Barrel
MAX0152NP20 20 L Plastic Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. MAX01521 Supplier MilliporeSigma Supplier No. AX01521
Add to Cart
Add to Cart

Recommended Storage

Store at room temperature

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 75-05-8
Molecular Formula C2H3N
Molecular Weight (g/mol) 41.053
InChI Key WEVYAHXRMPXWCK-UHFFFAOYSA-N
Synonym methyl cyanide, cyanomethane, ethanenitrile, methanecarbonitrile, ethyl nitrile, methane, cyano, acetonitrile, cyanure de methyl, methylkyanid, mecn
PubChem CID 6342
ChEBI CHEBI:38472
IUPAC Name acetonitrile
SMILES CC#N

Specifications

Melting Point -45.7°C
Quantity 4 L
Flash Point 2°C
Packaging Plastic Barrel
Physical Form Clear Liquid, Free From Particulates
Vapor Pressure 97 hPa (20°C)
Water 10ppm
Boiling Point 81.6°C (1013 hPa)
Formula Weight 41.05
Percent Purity 0.999
EINECSNumber 200-835-2
RTECSNumber AL7700000
Recommended Storage Store at room temperature
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.