Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Acetonitrile, for applications requiring extremely low water content, meets UV Specifications, ≥99.9%, Solstice

Catalog No. 6000276 Shop All Honeywell Research Chemicals Products
Change view
Click to view available options
Quantity:
20 L
Packaging:
HDPE Nowpak™
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
60-002-76 20 L HDPE Nowpak™
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 60-002-76 Supplier Solstice Supplier No. NP01720

Chemical Identifiers

CAS 75-05-8
Molecular Formula C2H3N
Molecular Weight (g/mol) 41.053
InChI Key WEVYAHXRMPXWCK-UHFFFAOYSA-N
Synonym methyl cyanide, cyanomethane, ethanenitrile, methanecarbonitrile, ethyl nitrile, methane, cyano, acetonitrile, cyanure de methyl, methylkyanid, mecn
PubChem CID 6342
ChEBI CHEBI:38472
IUPAC Name acetonitrile
SMILES CC#N

Specifications

CAS Max % 100.0000%
Density 0.7822 g/cm3
Quantity 20 L
Flash Point 6°C
Packaging HDPE Nowpak™
UN Number UN1648
Beilstein 741857
Boiling Point 82°C
Formula Weight 41
Percent Purity >99.9%
Chemical Name or Material Acetonitrile, B&J Brand™, for applications requiring extremely low water content, meets UV Specifications, ≥99.9%
Show More Show Less
ShelfLife 547 days from date of manufacture
DOTInformation Transport Hazard Class: 3; Packing Group:3: II; Proper Shipping Name: Acetonitrile
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.