Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Acetonitrile, B&J Brand™ LC-MS, for LC-MS and HPLC, >99.9%, Honeywell Burdick & Jackson
BIOPHARMA

Catalog No. p-7112032 Shop All Honeywell Research Chemicals Products
Click to view available options
Quantity:
4 L
4 x 2.5 L
6 x 1 L
Packaging:
GL45 Glass Bottle
Glass Bottle

Chemical Identifiers

CAS 75-05-8
Molecular Formula C2H3N
Molecular Weight (g/mol) 41.053
InChI Key WEVYAHXRMPXWCK-UHFFFAOYSA-N
Synonym methyl cyanide, cyanomethane, ethanenitrile, methanecarbonitrile, ethyl nitrile, methane, cyano, acetonitrile, cyanure de methyl, methylkyanid, mecn
PubChem CID 6342
ChEBI CHEBI:38472
IUPAC Name acetonitrile
SMILES CC#N

Specifications

CAS Max % 100.0000%
Density 0.7822 g/cm3
Boiling Point 82°C
Quantity 4 L
Flash Point 6°C
Packaging Glass Bottle
UN Number UN1648
Beilstein 741857
Water 0.01% max.
Solubility Information Miscible in Water at 20°C
Formula Weight 41
Percent Purity >99.9%
Grade LC-MS
Chemical Name or Material Acetonitrile, B&J Brand™ LC-MS, for LC-MS and HPLC, >99.9%
Show More Show Less
ShelfLife 547 days from date of manufacture
DOTInformation Transport Hazard Class: 3; Packing Group:3: II; Proper Shipping Name: Acetonitrile
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.