Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Acetonitrile, CHROMASOLV™, for HPLC, gradient grade, ≥99.9%, Solstice
BIOPHARMA

Catalog No. 6000243 Shop All Honeywell Research Chemicals Products
Change view
Click to view available options
Quantity:
100 mL
1 L
2.5 L
4 x 2.5 L
4 x 4 L
6 x 1 L
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
60-002-43 2.5 L
60-002-42 1 L
60-002-46 6 x 1 L
60-002-47 4 x 4 L
60-002-40 100 mL
60-046-486 4 x 2.5 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
Catalog No. 60-002-43 Supplier Solstice Supplier No. 348514X2.5L
Add to Cart
Add to Cart

CHROMASOLV™, gradient grade, for HPLC,≥99.9%

  • Solstice has over 200 years of history in producing high-purity chemicals
  • It is the leading brand offering High-Purity Solvents for dedicated applications.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 75-05-8
Molecular Formula C2H3N
Molecular Weight (g/mol) 41.053
MDL Number MFCD00001878
InChI Key WEVYAHXRMPXWCK-UHFFFAOYSA-N
Synonym methyl cyanide, cyanomethane, ethanenitrile, methanecarbonitrile, ethyl nitrile, methane, cyano, acetonitrile, cyanure de methyl, methylkyanid, mecn
PubChem CID 6342
ChEBI CHEBI:38472
IUPAC Name acetonitrile
SMILES CC#N

Specifications

Melting Point -48°C
Absorbance HPLC-gradient/254nm ≤0.5mAU
Density 0.786g/mL at 25°C
Quantity 2.5 L
Refractive Index n20/D 1.344
Vapor Pressure 72.8mmHg (20°C)
Linear Formula CH3CN
Beilstein 741857
Boiling Point 81°C to 82°C
Formula Weight 41.05g/mol
Percent Purity ≥99.9%
Grade CHROMASOLV™ Gradient
Chemical Name or Material Acetonitrile, for HPLC
Show More Show Less
Hazard Statement P210-P280-P305 + P351 + P338
Precautionary Statement H225-H302 + H312 + H332-H319
EINECSNumber 200-835-2
RTECSNumber AL7700000
Recommended Storage RT
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.