Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Acetonitrile CHROMASOLV™ Plus, for HPLC, ≥99.9%, Solstice

Catalog No. 6000231 Shop All Honeywell Research Chemicals Products
Change view
Click to view available options
Quantity:
1 L
2.5 L
4 x 2.5 L
4 x 4 L
6 x 1 L
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
60-002-31 1 L
60-002-34 6 x 1 L
60-002-33 4 x 2.5 L
60-002-35 4 x 4 L
60-002-32 2.5 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Catalog No. 60-002-31 Supplier Solstice Supplier No. 349981L
Add to Cart
Add to Cart

New Quality CHROMASOLV™ Plus Acetonitrile is suitable for all applications including:Mass spectrometry, HPLC, TLC, GPC, UV, IR, GC, Pesticide Residue Analysis, and RNA/DNA Synthesis and Purification. Product filtered through a 0.2μm filter.

  • For HPLC, spectrophotometry, and environmental analysis
  • Tested for HPLC-suitability via gradient analysis and UV-VIS spectroscopy
  • Low UV impurity profile
  • Low impurity gradient LC baseline
  • Very low levels of non-volatile impurities
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 75-05-8
Molecular Formula C2H3N
Molecular Weight (g/mol) 41.053
MDL Number MFCD00001878
InChI Key WEVYAHXRMPXWCK-UHFFFAOYSA-N
Synonym methyl cyanide, cyanomethane, ethanenitrile, methanecarbonitrile, ethyl nitrile, methane, cyano, acetonitrile, cyanure de methyl, methylkyanid, mecn
PubChem CID 6342
ChEBI CHEBI:38472
IUPAC Name acetonitrile
SMILES CC#N

Specifications

Melting Point -48°C
Absorbance HPLC-gradient/210nm ≤1mAU
Density 0.786g/mL at 25°C
Quantity 1 L
Refractive Index n20/D 1.344
Linear Formula CH3CN
Vapor Pressure 72.8mmHg (20°C)
Beilstein 741857
Boiling Point 81°C to 82°C
Formula Weight 41.05g/mol
Percent Purity ≥99.9%
Grade CHROMASOLV™ Plus
Chemical Name or Material Acetonitrile, for HPLC
Show More Show Less
Hazard Statement P210-P280-P305 + P351 + P338
Precautionary Statement H225-H302 + H312 + H332-H319
EINECSNumber 200-835-2
RTECSNumber AL7700000
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.