Learn More
Acetonitrile, Environmental Grade, 99.7+%
Description
Acetonitrile is utilized as a polar aprotic solvent in organic synthesis and in the purification of butadiene. Aqueous two-phase systems based on acetonitrile and carbohydrates play an important character in the extraction and purification of biomolecules called vanillins.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Specifications
| Melting Point | °C to 48°C to°C to 45°C |
| Color | Colorless |
| Appearance | Colorless liquid |
| Density | 0.786 |
| Quantity | 4 L |
| Flash Point | 5°C (41°F) |
| Ignition Point | 525°C (977°F) |
| Odor | Pungent |
| Physical Form | Liquid |
| Refractive Index | 1.344 |
| Show More |
Safety and Handling
| Hazard Category | H225-H302+H312+H332-H319 |
| Hazard Statement | Keep away from sources of ignition - No Smoking. Wear suitable protective clothing and gloves. |
| Precautionary Statement | P210-P233-P240-P241-P242-P243-P260-P264b-P270-P271-P280-P301+P312-P303+P361+P353-P304+P340-P305+P351+P338-P312-P314-P330-P363-P370+P378q-P501c |
| DOTInformation | DOT Class: 3, Packaging Group: II |
| EINECSNumber | 200-835-2 |
| RTECSNumber | AL7700000 |
| TSCA | Yes |
| Recommended Storage | Ambient temperatures |
RUO – Research Use Only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.