Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Acetonitrile (HPLC), Fisher Bioreagents™

Catalog No. BP24051 Shop All Fisherbrand Products
Click to view available options
Quantity:
1 L
4 L
Packaging:
Safe-Cote™ Amber Glass 
Wide Neck Amber Bottle

Submicron Filtered

  • Also available in recyclable FisherPak™ and NOWPak™ containers.
  • EcoSafPak™ is an environmentally friendly packaging system made of 100% recyclable material by an SFI certified supplier.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 75-05-8
Molecular Formula C2H3N
Molecular Weight (g/mol) 41.053
InChI Key WEVYAHXRMPXWCK-UHFFFAOYSA-N
Synonym methyl cyanide, cyanomethane, ethanenitrile, methanecarbonitrile, ethyl nitrile, methane, cyano, acetonitrile, cyanure de methyl, methylkyanid, mecn
PubChem CID 6342
ChEBI CHEBI:38472
IUPAC Name acetonitrile
SMILES CC#N

Specifications

Melting Point -46°C
Absorbance 0.01 AU max. at 254nm, 0.02 AU max. at 220nm, 0.04 AU max. at 210nm, 0.05 AU max. at 205nm, 0.07 AU max. at 200nm, 0.15 AU max. at 195nm, 1.00 AU max. at 190nm
Color Colorless
Decomposition Information >500°C
Density 0.781g/cm³
Boiling Point 81°C
Quantity 1 L
Identification Pass Test
Packaging Wide Neck Amber Bottle
Physical Form Liquid
Refractive Index 1.3405 to 1.3425 (at 25°C)
Residue after Evaporation 1.0ppm max.
Assay Percent Range ≥99.9 %
Titratable Acid 0.008mEq/g max.
Linear Formula CH3CN
Titratable Base 0.0006mEq/g max.
Vapor Pressure 97mbar at 20°C
Viscosity 0.36 cP at 20°C
Water 0.01% max.
Merck Index 15, 69
Formula Weight 41.05
Percent Purity ≥99.9%
Grade Also meets ACS specifications
Chemical Name or Material Acetonitrile
Show More Show Less
Hazard Statement DANGER!
Precautionary Statement Emergency Overview
Flammable liquid and vapor. Irritating to eyes. May cause skin and respiratory tract irritation. May cause central nervous system effects. Liver and kidney injuries may occur. Use personal protective equipment. Use only under a chemical fume hood. Keep away from open flames, hot surfaces and sources of ignition. Wash off immediately with plenty of water for at least 15 minutes. Immediate medical attention is required. Rinse immediately with plenty of water, also under the eyelids, for at least 15 minutes. Immediate medical attention is required. Immediate medical attention is required. Move to fresh air.

NFPA

Health:2
Flammability:3
Instability:0
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.