Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Acetonitrile, for HPLC

Catalog No. ac610010040
Click to view available options
Quantity:
4 L
Packaging:
EcoSafPak™

CAS: 75-05-8 | C2H3N | 41.053 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

For HPLC
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 75-05-8
Molecular Formula C2H3N
Molecular Weight (g/mol) 41.053
MDL Number MFCD00001878
InChI Key WEVYAHXRMPXWCK-UHFFFAOYSA-N
Synonym methyl cyanide, cyanomethane, ethanenitrile, methanecarbonitrile, ethyl nitrile, methane, cyano, acetonitrile, cyanure de methyl, methylkyanid, mecn
PubChem CID 6342
ChEBI CHEBI:38472
IUPAC Name acetonitrile
SMILES CC#N

Specifications

Melting Point -46°C
Absorbance (1 cm cell vs water), 0.01 max. at 254nm, 0.02 max. at 220nm, 0.04 max. at 210nm, 0.05 max. at 205nm, 0.07 max. at 200nm, 0.15 max. at 195nm, 1 max. at 190nm
Decomposition Information >500°C
Density 0.7810g/mL
Boiling Point 81°C to 82°C
Quantity 4 L
Flash Point 2°C (35.8°F)
Identification Pass Test
Packaging EcoSafPak™
Refractive Index 1.3405 - 1.3425
Residue after Evaporation 1ppm max.
Titratable Acid 0.008 meq/g max.
Titratable Base 0.0006 meq/g max.
Linear Formula CH3CN
Vapor Pressure Vapor Pressure: 97mbar at 20°C
UN Number 1648; Colorless
Beilstein 02, 183
Water 0.01% max.
Fieser 02,13; 12,450; 13,156; 15,1
Merck Index 15, 69
Formula Weight 41.04
Percent Purity ≥99.9%
Grade HPLC
Chemical Name or Material Acetonitrile, 99.9%
Show More Show Less
Hazard Category Danger
Hazard Statement HYGROSCOPIC

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.