Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Acetonitrile, LC/MS grade

Catalog No. ac615140025
Click to view available options
Quantity:
2.5 L
Packaging:
Glass Bottle

CAS: 75-05-8 | C2H3N | 41.053 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

ICP-OES/-MS standards
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 75-05-8
Molecular Formula C2H3N
Molecular Weight (g/mol) 41.053
MDL Number MFCD00001878
InChI Key WEVYAHXRMPXWCK-UHFFFAOYSA-N
Synonym methyl cyanide, cyanomethane, ethanenitrile, methanecarbonitrile, ethyl nitrile, methane, cyano, acetonitrile, cyanure de methyl, methylkyanid, mecn
PubChem CID 6342
ChEBI CHEBI:38472
IUPAC Name acetonitrile
SMILES CC#N

Specifications

Melting Point -46°C
Absorbance 0.005 AU max. at 254nm, 0.005 AU max. at 280nm, 0.01 AU max. at 230nm, 0.015 AU max. at 220nm, 0.015 AU max. at 225nm, 0.025 AU max. at 215nm, 0.03 AU max. at 210nm, 0.04 AU max. at 205nm, 0.05 AU max. at 200nm, 0.15 AU max. at 195nm, 1.00 AU max. at 190nm
Decomposition Information >500°C
Density 0.7810g/mL
Boiling Point 81°C to 82°C
Quantity 2.5 L
Flash Point 2°C (35.8°F)
Identification Pass Test
Packaging Glass Bottle
Residue after Evaporation 0.8ppm max.
Titratable Acid 0.008 meq/g max.
Titratable Base 0.0006 meq/g max.
Linear Formula CH3CN
Vapor Pressure Vapor Pressure: 97mbar at 20°C
UN Number 1648; Colorless
Water 0.01% max.
Merck Index 15, 69
Formula Weight 41.04
Percent Purity ≥99.9%
Grade LC-MS
Chemical Name or Material Acetonitrile
Show More Show Less
Hazard Category Danger
Hazard Statement GHS H Statement:
Causes serious eye irritation.
Harmful if inhaled.
Harmful if swallowed.
Harmful in contact with skin.
Highly flammable liquid and vapor.
Precautionary Statement GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Ground/Bond container and receiving equipment.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.