Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Acetonitrile, Optima™, for HPLC and GC, Fisher Chemical™ PROMO

Catalog No. A9964FB25 Shop All Fisherbrand Products
Click to view available options
:
4 L

High purity and versatile solvent suitable for HPLC, spectrophotometry, and pesticide residue analysis.

  • Screened for fluorescing contaminants, and UV-absorbing contaminants in the 190 to 254 nm range, to reduce interferences for smoother baselines
  • Tested for HPLC gradient suitability
  • Tested to ensure minimum halogenated residue levels (≤10 ng/L) to demonstrate suitability for pesticide analysis
  • Also meets ACS specifications
  • Intended for research use only
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 75-05-8
Molecular Formula C2H3N
Molecular Weight (g/mol) 41.053 g/mol
MDL Number MFCD00001878
InChI Key WEVYAHXRMPXWCK-UHFFFAOYSA-N
Synonym methyl cyanide, cyanomethane, ethanenitrile, methanecarbonitrile, ethyl nitrile, methane, cyano, acetonitrile, cyanure de methyl, methylkyanid, mecn
PubChem CID 6342
ChEBI CHEBI:38472
IUPAC Name acetonitrile
SMILES CC#N

Specifications

Melting Point -46°C
Absorbance 0.005 AU Max at 254 nm, 0.02 AU max at 220 nm, 0.04 AU max at 210 nm, 0.05 AU max at 205 nm, 0.07 AU max at 200 nm, 0.15 AU max at 195 nm, 1.00 AU max at 190 nm
Color Colorless
Decomposition Information >500°C
Density 0.781 g/cm3
Boiling Point 81°C
Quantity 4 L
Flash Point 49°C (120.2°F)
Identification Pass Test
Packaging Amber Soda Lime Glass
Physical Form Liquid
Refractive Index 1.3405 to 1.3425 (at 25°C)
Assay Percent Range Assay: 99.9% min by GC
Residue after Evaporation 0.8 ppm Max
Titratable Acid 0.008 mEq/g Max
Titratable Base 0.0006 mEq/g Max
Vapor Pressure 97 mbar at 20°C
UN Number 1648; Colorless
Viscosity 0.36 cP at 20°C
Water 0.01% Max
Formula Weight 41.05 amu
Percent Purity ≥99.9%
Grade High Purity Mobile Phase for HPLC and/or Extraction Solvent for GC Applications
Chemical Name or Material Acetonitrile
Show More Show Less
Hazard Statement DANGER!
Precautionary Statement

Emergency Overview: Flammable liquid and vapor. Irritating to eyes. May cause skin and respiratory tract irritation. May cause central nervous system effects. Liver and kidney injuries may occur. Use personal protective equipment. Use only under a chemical fume hood. Keep away from open flames, hot surfaces and sources of ignition. Wash off immediately with plenty of water for at least 15 minutes. Immediate medical attention is required. Rinse immediately with plenty of water, also under the eyelids, for at least 15 minutes. Immediate medical attention is required. Immediate medical attention is required. Move to fresh air.

NFPA

Health:2

Flammability:3

Instability:0

DOTInformation DOT Class 3, : Flammable Liquid
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.