Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Acetonitrile, puriss. p.a., ACS Reagent, Reag. Ph. Eur., ≥99.5% (GC), Solstice

Catalog No. 60046177 Shop All Honeywell Research Chemicals Products
Change view
Click to view available options
Quantity:
1 L
2.5 L
4 x 2.5 L
6 x 1 L
155 kg
Packaging:
Glass Bottle
Metal Drum
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
60-046-177 2.5 L Glass Bottle
60-046-176 1 L Glass Bottle
60-046-180 6 x 1 L Glass Bottle
60-046-179 4 x 2.5 L Glass Bottle
60-046-175 155 kg Metal Drum
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Catalog No. 60-046-177 Supplier Solstice Supplier No. 330192.5L
Add to Cart
Add to Cart

Puriss. p.a., ACS Reagent, Reag. Ph. Eur.,≥99.5% (GC)

ACS GENERAL USE SOLVENTS

  • value priced solution for applications such as preparative-scale HPLC, cleaning and organic synthesis
  • suitable for less demanding applications such as routine isocratic HPLC
  • variations and decrease issues traceable to solvent contamination
  • all solvents are tested to meet ACS general use specifications
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 75-05-8
Molecular Formula C2H3N
Molecular Weight (g/mol) 41.053
MDL Number MFCD00001878
InChI Key WEVYAHXRMPXWCK-UHFFFAOYSA-N
Synonym methyl cyanide, cyanomethane, ethanenitrile, methanecarbonitrile, ethyl nitrile, methane, cyano, acetonitrile, cyanure de methyl, methylkyanid, mecn
PubChem CID 6342
ChEBI CHEBI:38472
IUPAC Name acetonitrile
SMILES CC#N

Specifications

Melting Point -48°C
Acidity Free acid (as CH3COOH): ≤0.001%
Alkalinity Free alkali (as NH3): ≤0.001%
Density 0.786g/mL at 25°C
Quantity 2.5 L
Explosion Limit 0.16
Flash Point 2°C (35.6°F)
Ignition Point 973°F
Packaging Glass Bottle
Refractive Index n20/D 1.3435 to 1.3445
Vapor Pressure 72.8mmHg (20°C)
Linear Formula CH3CN
UN Number UN1648
Beilstein 741857
Boiling Point 82°C
Formula Weight 41.05g/mol
Percent Purity ≥99.5% (GC)
Grade ACS/puriss. p.a.
Chemical Name or Material Acetonitrile, Reag. Ph. Eur.
Show More Show Less
Hazard Category Danger
Hazard Statement P210-P280-P305 + P351 + P338
Precautionary Statement H225-H302 + H312 + H332-H319
DOTInformation 3/PGII
EINECSNumber 200-835-2
RTECSNumber AL7700000
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.