Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Acetophenone, ≥99.5% (GC), MilliporeSigma™ Supelco™

Catalog No. p-7187690 Shop All MilliporeSigma Supelco Products
Click to view available options
Quantity:
1 mL
5 mL

Acetophenone is a colourless liquid with a sweet pungent taste. It can be used in perfume industry, as a fragrance ingredient and also as a flavouring agent in food industries.

Cleaning Products, Cosmetics, Environmental, Food and Beverages and Personal Care
TRUSTED_SUSTAINABILITY

Specifications

Ignition Point 1058°F
CAS 98-86-2
Vapor Pressure 1 mmHg (15°C)
Vapor Density 4.1 (vs. air)
Density 1.03 g/mL (at 25°C)
Flash Point 76°C
Physical Form Neat
Linear Formula CH3 COC6H5
MDL Number MFCD00008724
Melting Point 19°C to 20°C
Boiling Point 202°C
Synonym Methyl phenyl ketone
Type Analytical Standard
Quantity 1 mL
Formula Weight 120.15
Percent Purity ≥99.5% (GC)
Grade Analytical Standard
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.