Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Acetyl bromide, 98%

Catalog No. p-7020503
Click to view available options
Quantity:
25 g
100 g
500 g

Catalog No. AAA1248514

Supplier: Thermo Scientific Chemicals A1248514

Only null left
Add to Cart

CAS: 506-96-7 | C2H3BrO | 122.949 g/mol

Acetyl Bromide is used as an acetylating agent in the synthesis of fine chemicals, agrochemicals and pharmaceuticals. It is also used as an intermediate for dyes. Further, it reacts with alcohols and amines to produce acetate esters and acetamides. In addition to this, it is involved in the synthetic process of the anti-HIV agent.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Acetyl Bromide is used as an acetylating agent in the synthesis of fine chemicals, agrochemicals and pharmaceuticals. It is also used as an intermediate for dyes. Further, it reacts with alcohols and amines to produce acetate esters and acetamides. In addition to this, it is involved in the synthetic process of the anti-HIV agent.

Solubility
Miscible with ether, chloroform, benzene and acetone.

Notes
Incompatible with strong oxidizing agents, strong bases and alcohols
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 506-96-7
Molecular Formula C2H3BrO
Molecular Weight (g/mol) 122.949
MDL Number MFCD00000114
InChI Key FXXACINHVKSMDR-UHFFFAOYSA-N
Synonym ethanoyl bromide, acetylbromide, acetic acid, bromide, ch3cobr, unii-o18v5xyo0g, hsdb 663, o18v5xyo0g, bromideacetyl, acbr, acetyl bromide
PubChem CID 10482
IUPAC Name acetyl bromide
SMILES CC(=O)Br

Specifications

Melting Point -96°C
Density 1.66
Boiling Point 75°C to 77°C
Flash Point >75°C (167°F)
Odor Acrid
Assay Percent Range ≥98%
Refractive Index 1.45
Linear Formula CH3COBr
Quantity 25 g
UN Number UN1716
Beilstein 1697546
Sensitivity Moisture sensitive
Merck Index 14,82
Solubility Information Miscible with ether,chloroform,benzene and acetone.
Formula Weight 122.95
Percent Purity ≥98%
Physical Form Liquid
Chemical Name or Material Acetyl bromide
Show More Show Less
Hazard Category H227-H314
Hazard Statement GHS H Statement
H314-H318-H227
Causes severe skin burns and eye damage.
Causes serious eye damage.
Combustible liquid.
Precautionary Statement P210-P260-P264b-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P370+P378q-P501c
DOTInformation Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: ACETYL BROMIDE
EINECSNumber 208-061-7
RTECSNumber AO5955000
TSCA Yes
storageNote1 Moisture Sensitive
Recommended Storage Ambient temperatures; Store under Nitrogen

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.