Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Acetyl β-Catenin (Lys49) Rat anti-Human, Clone: mAb-K49-Ac, MilliporeSigma™

Catalog No. MABE182925 Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μg
25 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MABE182925 25 μg
MABE1829100 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. MABE182925 Supplier MilliporeSigma Supplier No. MABE182925UG
Add to Cart
Add to Cart

Rat Monoclonal Antibody

Acetyl β-Catenin (Lys49) Monoclonal antibody specifically detects Acetyl β-Catenin (Lys49) in Human samples. It is validated for Dot Blot, Immunoprecipitation, Western Blotting.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Acetyl β-Catenin (Lys49)
Applications Dot Blot, Immunoprecipitation, Western Blot
Classification Monoclonal
Clone mAb-K49-Ac
Conjugate Unconjugated
Formulation Purified rat monoclonal antibody IgG2b in buffer containing 0.1 M Tris-Glycine (pH 7.4), 150 mM NaCl with 0.05% sodium azide.
Host Species Rat
Immunogen BSA-conjugated linear peptide corresponding to 19 amino acids surrounding acetylated lysine 49 from human -catenin.
Purification Method Protein G purified
Quantity 25 μg
Regulatory Status RUO
Primary or Secondary Primary
Target Species Human
Content And Storage Stable for 1 year at 2-8°C from date of receipt.
Form Purified
Isotype IgG2b κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.