Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Acetyl chloride, 98%, ACROS Organics™

Click to view available options
Quantity:
1L
2.5L
Packaging:
Glass bottle
75-36-5
C2H3ClO
78.5
WETWJCDKMRHUPV-UHFFFAOYSA-N
ethanoyl chloride,acetic chloride,acetylchloride,acetic acid chloride,ch3cocl,rcra waste number u006,acetic acid, chloride,unii-qd15rno45k,ccris 4568,hsdb 662
6367
CHEBI:37580
acetyl chloride
CC(=O)Cl
75-36-5
C2H3ClO
78.5
WETWJCDKMRHUPV-UHFFFAOYSA-N
ethanoyl chloride,acetic chloride,acetylchloride,acetic acid chloride,ch3cocl,rcra waste number u006,acetic acid, chloride,unii-qd15rno45k,ccris 4568,hsdb 662
6367
CHEBI:37580
acetyl chloride
CC(=O)Cl
Boiling Point 51.0°C
Refractive Index 1.3876 to 1.3896
Color Undesignated
Density 1.1000g/mL
Infrared Spectrum Authentic
Melting Point -112.0°C
Quantity 2.5L
Specific Gravity 1.1
Packaging Glass bottle
Flash Point 4°C
Assay Percent Range 98%
Linear Formula CH3COCl
Beilstein 02, 173
Fieser 01,11; 04,5; 05,4; 07,3; 11,4
Merck Index 15, 80
Solubility Information Solubility in water: reacts. Other solubilities: miscible with acetone, benzenz, toluene,, chloroform, ether, petroleum ether, carbon dosulfi, de and glacial acetic acid, soluble in ccl4, reacts with alcohol
Formula Weight 78.5
Physical Form Liquid
Percent Purity 98%
Chemical Name or Material Acetyl chloride
Show More Show Less