Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Acetyl chloride, 99+%

Catalog No. ac219470250
Click to view available options
Quantity:
25 mL
250 mL
1 L
2.5 L
Packaging:
Glass bottle
This item is not returnable. View return policy
This item is not returnable. View return policy

Acetyl chloride, 99+%, C2H3ClO, CAS Number-75-36-5, acetic acid chloride, acetic acid, chloride, acetic chloride, acetylchloride, ccris 4568, ch3cocl, ethanoyl chloride, hsdb 662, rcra waste number u006, unii-qd15rno45k, 250mL, 99% min. (GC), 02, 173, 51.0 deg.C, 100.0, 99.0, CHEBI:37580 | CAS: 75-36-5 | C2H3ClO | 78.50 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 75-36-5
Molecular Formula C2H3ClO
Molecular Weight (g/mol) 78.50
MDL Number MFCD00000719
InChI Key WETWJCDKMRHUPV-UHFFFAOYSA-N
Synonym ethanoyl chloride, acetic chloride, acetylchloride, acetic acid chloride, ch3cocl, rcra waste number u006, acetic acid, chloride, unii-qd15rno45k, ccris 4568, hsdb 662
PubChem CID 6367
ChEBI CHEBI:37580
IUPAC Name acetyl chloride
SMILES CC(Cl)=O

Specifications

CAS Min % 99.0
CAS Max % 100.0
Melting Point -112.0°C
Density 1.1000g/mL
Boiling Point 51.0°C
Flash Point 4°C
Infrared Spectrum Authentic
Assay Percent Range 99% min. (GC)
Packaging Glass bottle
Refractive Index 1.3876 to 1.3896
Linear Formula CH3COCl
Quantity 25 mL
Beilstein 02, 173
Fieser 01,11; 04,5; 05,4; 07,3; 11,4
Specific Gravity 1.1
Merck Index 15, 80
Solubility Information Solubility in water: reacts. Other solubilities: miscible with acetone, benzenz, toluene,, chloroform, ether, petroleum ether, carbon dosulfi, de and glacial acetic acid, soluble in ccl4, reacts with alcohol
Formula Weight 78.5
Percent Purity 99+%
Physical Form Liquid
Chemical Name or Material Acetyl chloride, p.a.
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement
Highly flammable liquid and vapor.
Causes severe skin burns and eye damage.
Reacts violently with water.
Corrosive to the respiratory tract.
Precautionary Statement GHS P Statement
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN (or hair): Take off immediately all contaminated clothing.
Rin
EINECSNumber 200-865-6
RTECSNumber AO6390000
TSCA TSCA

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.