Acetyl chloride, 99+%, Alfa Aesar™

Manufacturer: Alfa Aesar 43262K2

Quantity 1L

 View more versions of this product

Catalog No. AA43262K2

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 75-36-5
    Molecular Formula C2H3ClO
    Linear Formula CH3COCl
    Formula Weight 78.5
    MDL Number MFCD00000719
    UN Number UN1717
    Beilstein 605303
    Merck Index 14,85
    Synonym Ethanoyl chloride; Acetic chloride
    Chemical Name or Material Acetyl chloride

    Specifications

    Formula Weight 78.5
    Quantity 1L
    UN Number UN1717
    Merck Index 14,85
    Density 1.104
    Health Hazard 1 Danger
    Melting Point −112°C
    Flash Point 4°C (39°F)
    DOT Information Transport Hazard Class: 3; Packing Group: II; Proper Shipping Name: ACETYL CHLORIDE
    RTECS Number AO6390000
    Physical Form Liquid
    Molecular Formula C2H3ClO
    MDL Number MFCD00000719
    Beilstein 605303
    Synonym Ethanoyl chloride; Acetic chloride
    Boiling Point 50° to 52°C
    Health Hazard 2 GHS H Statement
    H225-H314;
    Highly flammable liquid and vapor.
    Causes severe skin burns and eye damage.
    TSCA TSCA
    Refractive Index 1.3890
    Linear Formula CH3COCl
    CAS 75-36-5
    Color Colorless
    Odor Pungent
    Sensitivity Moisture Sensitive
    Health Hazard 3 GHS P Statement
    P210-P260-P303+P361+P353-P305+P351+P338-P405-P501a;
    Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
    Do not breathe dust/fume/gas/mist/vapours/spray.
    IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.
    Chemical Name or Material Acetyl chloride
    Assay ≥99%

    GHS H Statement
    H225-H314;
    Highly flammable liquid and vapor.
    Causes severe skin burns and eye damage.

    GHS P Statement
    P210-P260-P303+P361+P353-P305+P351+P338-P405-P501a;
    Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
    Do not breathe dust/fume/gas/mist/vapours/spray.
    IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.

    Danger

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit