Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Acetyl CholinesteraseGoat anti-Bovine, Polyclonal, Rockland™
SDP

Catalog No. 501989985
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
2 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-198-9985 2 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-198-9985 Supplier Rockland Immunochemicals Supplier No. 100101232
Add to Cart
Add to Cart

Goat Polyclonal Antibody

Store vial at 4°C prior to restoration. For extended storage aliquot contents and freeze at -20°C or below. Avoid cycles of freezing and thawing. Centrifuge product if not completely clear after standing at room temperature. This product is stable for several weeks at 4°C as an undiluted liquid. Dilute only prior to immediate use. Assay by immunoelectrophoresis resulted in a single precipitin arc against anti-goat serum, purified and partially purified Acetyl Cholinesterase [Bovine Erythrocytes]. Cross reactivity against Acetyl Cholinesterase from other sources is unknown.

Acetylcholinesterase hydrolyzes the neurotransmitter, acetylcholine at neuromuscular junctions and brain cholinergic synapses, and thus terminates signal transmission. The Protein is also found on the red blood cell membranes, where it constitutes the Yt blood group antigen. Acetylcholinesterase exists in multiple molecular forms which possess similar catalytic properties, but differ in their oligomeric assembly and mode of cell attachment to the cell surface. The major form of acetylcholinesterase found in brain, muscle and other tissues is the hydrophilic species, which forms disulfide-linked oligomers with collagenous, or lipid-containing structural subunits.

Specifications

Antigen Acetyl Cholinesterase
Applications ELISA, Immunoprecipitation, Western Blot
Classification Polyclonal
Concentration 90 mg/mL
Conjugate Unconjugated
Formulation 0.02M potassium phosphate with 0.15M NaCl and 0.01% sodium azide; pH 7.2
Gene ACHE
Gene Accession No. P23795
Gene Alias ACE-1; ACEE; Acetylcholinesterase; acetylcholinesterase (Cartwright blood group); acetylcholinesterase (Yt blood group); Ache; apoptosis-related acetylcholinesterase; ARACHE; BOS_23015; EC 3.1.1.7; glycolipid-anchored form of acetylcholinesterase; N-ACHE; OTTHUMP00000211349; OTTHUMP00000211356; YT; Yt blood group
Gene Symbols ACHE
Host Species Goat
Immunogen Acetyl Cholinesterase [Bovine Erythrocytes]
Quantity 2 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 540446
Target Species Bovine
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Lyophilized
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.