Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Acetyl CoA Carboxylase 1(ACC1) Rabbit anti-Human, Clone: 2D3, Bioss
SDP

Catalog No. 501989688
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-198-9688 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-198-9688 Supplier Bioss Supplier No. BSM52486R
Add to Cart
Add to Cart

Rabbit Monoclonal Antibody

Acetyl-CoA carboxylase (ACC) is a complex multifunctional enzyme system. ACC is a biotin-containing enzyme which catalyzes the carboxylation of acetyl-CoA to malonyl-CoA, the rate-limiting step in fatty acid synthesis. There are two ACC forms, alpha and beta, encoded by two different genes. ACC-alpha is highly enriched in lipogenic tissues. The enzyme is under long term control at the transcriptional and translational levels and under short term regulation by the phosphorylation/dephosphorylation of targeted serine residues and by allosteric transformation by citrate or palmitoyl-CoA. Multiple alternatively spliced transcript variants divergent in the 5′ sequence and encoding distinct isoforms have been found for this gene.

Specifications

Antigen Acetyl CoA Carboxylase 1(ACC1)
Applications Immunohistochemistry, Western Blot
Classification Monoclonal
Clone 2D3
Concentration 1 μg/mL
Conjugate Unconjugated
Formulation TBS with 1% BSA, 40% glycerol and 0.05% sodium azide; pH
Gene Acaca
Gene Accession No. Q13085
Gene Alias A1Z784_DROME; A530025K05Rik; Acac; ACACA; ACACAD; ACC; ACC1; ACCA; ACC-alpha; ACC-PA; ACC-PB; ACC-PC; ACC-PE; ACC-PF; ACC-PG; aceto-acetyl-CoA-thiolase; acetyl CoA carboxylase; acetyl coenzyme A carboxylase; Acetyl-CoA carboxylase; Acetyl-CoA carboxylase 1; Acetyl-CoA carboxylase 265; acetyl-CoA carboxylase alpha; acetyl-coenzyme A carboxylase; Acetyl-Coenzyme A carboxylase alpha; ACoT; Biotin carboxylase; BOS_18465; CG11198; CG11198 gene product from transcript CG11198-RG; CG11198-PA; CG11198-PB; CG11198-PC; CG11198-PE; CG11198-PF; CG11198-PG; CG8723; dACC; DmACC; Dmel\CG11198; Dmel_CG11198; EC 6.3.4.14; EC 6.4.1.2; FBgn0043811; Gm738; I79_006999
Gene Symbols Acaca
Host Species Rabbit
Immunogen Recombinant human ACC1 between 350 to 450 amino acids.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 31
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.