Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Acetyl CoA Carboxylase 1 Mouse anti-Human, Clone: 10H2.1, MilliporeSigma™

Catalog No. MABS202425 Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μg
25 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MABS2024-25 25 μg
MABS2024-100 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. MABS2024-25 Supplier MilliporeSigma Supplier No. MABS202425UG
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Anti-Acetyl CoA Carboxylase 1, clone 10H2.1 is a mouse monoclonal antibody that detects Acetyl-CoA carboxylase 1 and has been tested for use in Western Blotting and Immunohistochemistry (Paraffin).
TRUSTED_SUSTAINABILITY

Specifications

Antigen Acetyl CoA Carboxylase 1
Applications Western Blot, Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone 10H2.1
Concentration Please refer to lot specific datasheet.
Conjugate Unconjugated
Gene Symbols ACACA,ACAC,ACC1,ACCA
Host Species Mouse
Immunogen GST-tagged recombinant fragment corresponding to 245 amino acids from the C-terminal region of human Acetyl-CoA carboxylase 1.
Purification Method Protein G Purified
Quantity 25 μg
Regulatory Status RUO
Research Discipline Signaling
Primary or Secondary Primary
Gene ID (Entrez) NP_942133
Test Specificity Clone 10H2.1 detects human Acetyl-CoA carboxylase 1. It targets an epitope within 245 amino acids from the C-terminal end.
Target Species Human
Content And Storage Stable for 1 year at 2-8°C from date of receipt.
Form Purified
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.