Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Acetyl CoA Carboxylase 1 Mouse anti-Mouse, Human, Clone: 7H4.2, MilliporeSigma™

Catalog No. MABS830MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MABS830MI 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MABS830MI Supplier MilliporeSigma Supplier No. MABS830
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Anti-Acetyl CoA Carboxylase 1 Antibody, clone 7H4.2 is a mouse monoclonal antibody that targets Acetyl CoA Carboxylase 1 and has been tested for use in Immunohistochemistry (Paraffin) and Western Blotting.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Acetyl CoA Carboxylase 1
Applications Western Blot, Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone 7H4.2
Concentration Please refer to lot specific datasheet.
Conjugate Unconjugated
Gene Symbols ACACA,ACAC,ACC1,ACCA
Host Species Mouse
Immunogen GST/His-tagged recombinant fragment corresponding to 86 amino acids from the C-terminal end of human Acetyl-CoA carboxylase 1.
Purification Method Protein G Purified
Quantity 100 μg
Regulatory Status RUO
Research Discipline Signaling
Primary or Secondary Primary
Gene ID (Entrez) NP_942133
Test Specificity Clone 7H4.2 detects acetyl-CoA carboxylase 1 in human. It targets an epitope within 86 amino acids from the C-terminal end.
Target Species Mouse, Human
Content And Storage Stable for 1 year at 2-8°C from date of receipt.
Form Purified
Isotype IgG2b κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.