Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Acetyl CoA Carboxylase 1, Rabbit anti-Human, Polyclonal, Millipore Sigma™

Catalog No. ABS106825UL Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
25 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ABS106825UL 25 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. ABS106825UL Supplier MilliporeSigma Supplier No. ABS106825UL
Only null left
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Anti-Acetyl CoA Carboxylase 1, Cat. No. ABS1068, is a rabbit polyclonal antibody that detects Acetyl CoA Carboxylase 1 and has been tested for use in Immunohistochemistry (Paraffin) and Western Blotting.

Acetyl-CoA carboxylase 1 (UniProt: Q13085; also known as EC: 6.4.1.2, ACC1, ACC-alpha) is encoded by the ACACA (also known as ACAC, ACC1, ACCA) gene (Gene ID: 31) in human. ACC1 is a cytoplasmic protein that is expressed in brain, placental, skeletal muscle, renal, pancreatic and adipose tissues. It is also expressed at low level in pulmonary tissue, but is not detected in the liver. ACC1 can be present as a monomer, homodimer, or homotetramer. It can also form filamentous polymers. Four isoforms of ACC1 have been described that are produced by alternative splicing. It catalyzes the rate-limiting reaction in the biogenesis of long-chain fatty acids and carries out three functions: biotin carboxyl carrier protein, biotin carboxylase and carboxyltransferase. The activity of ACC can be controlled at the transcriptional level as well as by small molecule modulators and covalent modification. ACC1 levels are regulated under long term control at the transcriptional and translational levels and under short term regulation by the phosphorylation/dephosphorylation of targeted serine residues and by allosteric transformation by citrate or palmitoyl-CoA. Mutations in ACACA gene can result in ACC1 deficiency and lead to error of de novo fatty acid synthesis associated with severe brain damage, persistent myopathy and poor growth.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Acetyl CoA Carboxylase 1
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Conjugate Unconjugated
Dilution Immunohistochemistry Analysis: A 1:50 dilution from a representative lot detected Acetyl CoA Carboxylase 1 in human liver and human kidney tissue sections.
Formulation Purified rabbit polyclonal antibody in buffer containing 0.1 M Tris-Glycine (pH 7.4), 150 mM NaCl with 0.05% sodium azide.
Gene Alias ACC1;ACC-alpha;EC 6.4.1.2
Gene Symbols ACACA;ACC1;ACCA
Host Species Rabbit
Immunogen GST-tagged recombinant fragment corresponding to 245 amino acids from the C-terminal half of human Acetyl-CoA carboxylase 1.
Molecular Weight of Antigen ∽220 kDa observed; 265.55 kDa calculated. Uncharacterized bands may be observed in some lysate(s).
Purification Method Affinity Purified
Quantity 25 μL
Regulatory Status RUO
Research Discipline Signaling
Primary or Secondary Primary
Test Specificity This rabbit polyclonal antibody detects Acetyl-CoA carboxylase 1 in human cells. It targets an epitope within 245 amino acids from the C-terminal region.
Target Species Human
Content And Storage Stable for 1 year at 2-8°C from date of receipt.
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.