Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Acetyl-CoA Carboxylase (ACC) Monoclonal Antibody (143), Abbomax

Catalog No. 502204209
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-220-4209 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-220-4209 Supplier Abbomax Inc Supplier No. 604030
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Positive control: K562 Cellular location: Cytoplasm.

Acetyl-CoA Carboxylase (ACC) Monoclonal antibody specifically detects Acetyl-CoA Carboxylase (ACC) in Human samples. It is validated for Immunocytochemistry, Immunohistochemistry (Paraffin), Western Blot

Specifications

Antigen Acetyl-CoA Carboxylase (ACC)
Applications Immunocytochemistry, Immunohistochemistry (Paraffin), Western Blot
Classification Monoclonal
Clone 143
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with proprietary stabilizer and 0.01% sodium azide
Gene Acaca
Gene Accession No. Q13085
Gene Alias A1Z784_DROME; A530025K05Rik; Acac; Acaca; ACACAD; Acc; ACC1; ACCA; ACC-alpha; ACC-PA; ACC-PB; ACC-PC; ACC-PE; ACC-PF; ACC-PG; aceto-acetyl-CoA-thiolase; acetyl CoA carboxylase; acetyl coenzyme A carboxylase; Acetyl-CoA carboxylase; Acetyl-CoA carboxylase 1; acetyl-CoA carboxylase 265; acetyl-CoA carboxylase alpha; acetyl-CoA-carboxylase; acetyl-coenzyme A carboxylase; acetyl-coenzyme A carboxylase alpha; ACoT; Biotin carboxylase; CG11198; CG11198-PA; CG11198-PB; CG11198-PC; CG11198-PE; CG11198-PF; CG11198-PG; CG8723; dACC; DmACC; Dmel\CG11198; Dmel_CG11198; EC 6.3.4.14; EC 6.4.1.2; FBgn0043811; GH12002p; Gm738; I79_006999; unnamed protein product
Host Species Mouse
Immunogen A synthetic peptide derived from C-terminus of Acetyl-CoA Carbvoxylase 1 protein
Purification Method Protein A
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 31
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.