Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Acetyl-CoA Carboxylase alpha/ACACA Antibody, Alexa Fluor™ 750, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NB10055246AF750
Description
Acetyl-CoA Carboxylase alpha/ACACA Polyclonal antibody specifically detects Acetyl-CoA Carboxylase alpha/ACACA in Human samples. It is validated for Western BlotSpecifications
Acetyl-CoA Carboxylase alpha/ACACA | |
Polyclonal | |
Western Blot | |
ACACacetyl-CoA carboxylase 1, ACC1ACC, ACCA, ACC-alpha, acetyl-CoA carboxylase alpha, acetyl-Coenzyme A carboxylase alpha, EC 6.4.1.2 | |
The immunogen recognized by this antibody maps to a region between residue 1925 and 1975 of human acetyl-Coenzyme A carboxylase alpha using the numbering given in entry NP_942131.1 (GeneID 31). | |
0.1 mL | |
Breast Cancer, Lipid and Metabolism, Neuroscience, Phospho Specific | |
31 | |
Store at 4C in the dark. | |
IgG |
Western Blot | |
Alexa Fluor 750 | |
50mM Sodium Borate | |
Rabbit | |
Immunogen affinity purified | |
RUO | |
Primary | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction