Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Acetyl-CoA Carboxylase alpha/ACACA, p Ser79 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP313324
This item is not returnable.
View return policy
Description
Acetyl-CoA Carboxylase alpha/ACACA Polyclonal antibody specifically detects Acetyl-CoA Carboxylase alpha/ACACA in Human, Mouse, Rat samples. It is validated for Western BlotSpecifications
| Acetyl-CoA Carboxylase alpha/ACACA | |
| Polyclonal | |
| PBS, 1% BSA, 20% Glycerol (pH7) | |
| Rabbit | |
| Antigen Affinity-purified | |
| RUO | |
| Primary | |
| Human, Mouse, Rat | |
| Purified |
| Western Blot | |
| Unconjugated | |
| ACACacetyl-CoA carboxylase 1, ACC1ACC, ACCA, ACC-alpha, acetyl-CoA carboxylase alpha, acetyl-Coenzyme A carboxylase alpha, EC 6.4.1.2 | |
| Carrier-protein conjugated synthetic peptide corresponding to residues around Rat Acetyl-CoA Carboxylase alpha/ACACA (phospho Ser79). The exact sequence is proprietary. | |
| 100 μg | |
| Breast Cancer, Lipid and Metabolism, Neuroscience, Phospho Specific | |
| 31 | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction