Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Acetyl-CoA Carboxylase alpha/ACACA, p Ser79 Antibody (SR1413), Novus Biologicals™

Rabbit Monoclonal Antibody
$302.50 - $423.00
Specifications
Antigen | Acetyl-CoA Carboxylase alpha/ACACA |
---|---|
Clone | SR1413 |
Dilution | Western Blot 1:500-1:2000 |
Classification | Monoclonal |
Conjugate | Unconjugated |
Description
Acetyl-CoA Carboxylase alpha/ACACA Monoclonal antibody specifically detects Acetyl-CoA Carboxylase alpha/ACACA in Human, Mouse, Rat samples. It is validated for Western BlotSpecifications
Acetyl-CoA Carboxylase alpha/ACACA | |
Western Blot 1:500-1:2000 | |
Unconjugated | |
Rabbit | |
Breast Cancer, Lipid and Metabolism, Neuroscience, Phospho Specific | |
PBS, pH 7.4, 150mM NaCl, 50% glycerol. | |
31 | |
Affinity purified |
SR1413 | |
Monoclonal | |
Purified | |
RUO | |
Human, Mouse, Rat | |
ACACacetyl-CoA carboxylase 1, ACC1ACC, ACCA, ACC-alpha, acetyl-CoA carboxylase alpha, acetyl-Coenzyme A carboxylase alpha, EC 6.4.1.2 | |
Primary | |
Store at -20° C. Avoid freeze/thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title